Learn More
Thermo Scientific Chemicals β-Alanine, 98%
Description
Effective catalyst for the Knoevenagel condensation. Beta Alanine is used as nutrition supplements in food production. Increase muscular strength & power output, Increases Muscle Mass, increase Anaerobic Endurance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Effective catalyst for the Knoevenagel condensation. Beta Alanine is used as nutrition supplements in food production. Increase muscular strength & power output, Increases Muscle Mass, increase Anaerobic Endurance.
Solubility
Soluble in water(550g/L). Slightly soluble in alcohol. Insoluble in ether and acetone.
Notes
Slightly hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | ∼205°C (decomposition) |
| Assay Percent Range | 98% |
| Linear Formula | H2NCH2CH2CO2H |
| Beilstein | 906793 |
| Merck Index | 14,205 |
| Solubility Information | Soluble in water(550g/L). Slightly soluble in alcohol. Insoluble in ether and acetone. |
| Density | 1.437 |
| Quantity | 5000 g |
| Odor | Odorless |
| Formula Weight | 89.09 |
| Show More |
Safety and Handling
| EINECSNumber | 203-536-5 |
| RTECSNumber | UA2369200 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.