Learn More
Bioassay Systems EnzyChrom™ Alanine Transaminase Assay Kit. For quantitative determination of alanine transaminase activity.
SDP
Catalog No. 50489244 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of alanine transaminase enzyme activity. Key Features: Sensitive. Linear detection range: 2-100 U/L. Simple and convenient. This simple, convenient assay can be carried out in a microplate or a cuvette and takes only 10 min. Method: OD340nm. Samples: Serum, plasma etc. Species: All. Procedure: Assay takes 10 min. Kit size: 100 tests. Detection limit: 2 U/L.
Catalog No. 50-489-244 Supplier Bioassay Systems Supplier No. EALT100
For quantitative determination of alanine transaminase enzyme activity. Key Features: Sensitive. Linear detection range: 2-100 U/L. Simple and convenient. This simple, convenient assay can be carried out in a microplate or a cuvette and takes only 10 min. Method: OD340nm. Samples: Serum, plasma etc. Species: All. Procedure: Assay takes 10 min. Kit size: 100 tests. Detection limit: 2 U/L.