Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alanine, USP, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18600340 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-340 25 g Amber Glass Bottle
18-600-341 100 g Amber Glass Bottle
18-600-342 1 kg Amber Glass Bottle
18-600-343 5 kg Poly Pail
18-600-344 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-600-340 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A125325GM

A1253, 56-41-7, C3H7NO2

Spectrum™ Chemical Alanine, USP is a non-essential alpha-amino acid and appears as a white powder. Alanine is used in clinical nutrition and as an ingredient for pharmaceutical products such as therapeutics for prostate hypertrophy. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 56-41-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064410
InChI Key QNAYBMKLOCPYGJ-REOHCLBHSA-N
IUPAC Name (2S)-2-aminopropanoic acid
SMILES C[C@H](N)C(O)=O

Specifications

CAS Max % 100%
Chloride 0.05%
Ignition Residue 0.15%
Iron (Fe) 30 ppm
Loss on Drying 0.2%
Packaging Amber Glass Bottle
Specific Rotation +13.7° to +15.1°
Sulfate 0.03%
Quantity 25 g
Percent Purity 98.5 to 101.5%
Grade USP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.