Learn More
Medchemexpress LLC Alantolactone | 546-43-0 | MFCD00274568 | 99.9% | 232.32 g/mol | C15H20O2 | 10 MG

Supplier: Medchemexpress LLC HYN003810MG
Alantolactone is a sesquiterpene lactone used in research as a selective STAT3 inhibitor with potent anticancer activity; it induces apoptosis in cancer cells and is supplied as a solid with characterized purity, solubility, and storage recommendations suitable for in vitro and in vivo studies.
- Selective STAT3 inhibitor suitable for cancer biology research.
- High purity (≈99.9%) supports reproducible results.
- Soluble in DMSO at high concentrations for in vitro assays.
- Available in small pack sizes (including 10 MG) for pilot studies.
- Recommended storage conditions provided for long-term stability.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.