Learn More
Selleck Chemical LLC Samotolisib
SDP
Supplier:  Selleck Chemical LLC S8322100MG
Encompass_Preferred
Samotolisib (LY3023414 GTPL8918) is an oral ATP competitive inhibitor of the class I PI3K isoforms mTOR and DNA-PK
Catalog No. 50-000-01090
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.