Learn More
Abcam Alaproclate hydrochloride, serotonin uptake inhibitor, 25MG
Supplier:  Abcam AB14591725MG
Encompass_Preferred
MW 292.2 Da, Purity >95%. Potent, selective serotonin uptake inhibitor. Potent, reversible, noncompetitive NMDA antagonist (IC₅₀ = 0.3 μM). Kv1.2 channel blocker (IC₅₀ = 6.9 μM). Shows antidepressant effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5014