Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALAS1 Monoclonal Antibody (OTI1C5), TrueMAB™, OriGene
SDP

Catalog No. 501681950
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-1950 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-1950 Supplier OriGene Supplier No. TA808177
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ALAS-1 is a nuclear-encoded mitochondrial matrix enzyme which catalyzes the condensation of glycine with succinyl-CoA to form delta-amino levulinate, CO2 and CoA. It regulates the first and rate-limiting step of heme biosynthetic pathway. It is one of the two isoforms of ALAS and is a pyridoxal-5′ phosphate dependant housekeeping enzyme. It is ubiquitously expressed and is responsible of providing heme for cytochromes and other hemoproteins. ALAS1 in liver undergoes negative feedback regulation by heme. Transcription of ALAS1 gene is stimulated by cAMP and respiratory uncoupling whereas Phorbol esters and insulin repress ALAS1 gene expression. ALAS1 is suggested to be involved in reciprocal regulation of Heme biosynthesis and circadian clock suggesting a potential target for treatment of circadian disorders. ALAS1 gene expression is down regulated in Acute Liver failure resulting in altered heme metabolism and liver function.

Specifications

Antigen ALAS1
Applications Western Blot
Classification Monoclonal
Clone OTI1C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ALAS1
Gene Accession No. P13196
Gene Alias ALAS, ALAS-H, ALAS3, ALASH, MIG4
Gene Symbols ALAS1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 57-308 of human ALAS1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 211
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.