Learn More
Description
Specifications
Specifications
| Cross Reactivity | Guinea pig |
| Target Species | Guinea Pig |
| For Use With (Application) | Quantitative |
| Gene ID (Entrez) | 100379232 |
| Quantity | 1 Kit |
| Regulatory Status | RUO |
| Storage Requirements | Store TMB Substrate, Wash Buffer (30X concentrate) and the Stop Solution at 4°C upon receipt while the others should be at -20°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.