Learn More
Dukal™ Albahealth™ Iodoform Packing Strips
Description
- Made of a woven gauze fabric which reduces linting and fraying.
- Iodoform provides odor-resistance to the packing strip.
- Unbreakable poly bottles.
- Tamper evident seals.
- Used in wound care management applications such as nasal or sinus packing.
Specifications
Specifications
| Product Type | Iodoform Packing Strips |
| Sterility | Sterile |
| Dimensions (L x W) | 5 yd. x 1 in. |
| Latex Free | Yes |