Learn More
Dukal™ Albahealth™ Oil Emulsion Dressings
Description
- Made of a permeable knitted fabric that allows wounds to drain freely into the absorbent dressing.
- Non-adherent petrolatum.
- Enhances wound drainage.
- Minimizes maceration.
- Supports healing environment.
- Unique sterile stay flat packaging.
- Ideal for draining surgical wounds, open ulcers, abrasions and first or second degree burns.
Specifications
Specifications
| Product Type | Oil Emulsion Dressing |
| Sterility | Sterile |
| Quantity | 36/Cs. |
| Dimensions (L x W) | 3 x 16 in. |
| Latex Free | Yes |