Learn More
Medchemexpress LLC Albiglutide fragment TFA | 99.1% | 3,283.60 g/mol (free base) | C148H224N40O45.xC2HF3O2 | 5 MG

Supplier: Medchemexpress LLC HY108795A5MG
Albiglutide fragment TFA is a synthetic GLP-1 (7-36) analog peptide fragment supplied as the trifluoroacetic acid (TFA) salt. It is provided as a lyophilized powder for research use in GLP-1 receptor biology and type 2 diabetes models. The peptide sequence and molecular weight (free base 3,283.60 g/mol) are documented in the product datasheet; handle and store under inert, dry conditions to preserve stability.
- Lyophilized peptide TFA salt, suitable for research.
- Amino acid sequence corresponds to GLP-1 (7-36) analog.
- Molecular weight 3,283.60 g/mol (free base).
- High purity for biological assays.
- Store frozen under inert, dry conditions; powder stable at -80°C up to 2 years.
- Supplied as a small-scale research quantity (5 MG).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.