Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Albite, naturally occurring mineral, grains, ≈0.06 to 0.19 in., Thermo Scientific™

Catalog No. AA4264103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4264103 1 g
AA4264106 5 g
AA4264114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4264103 Supplier Thermo Scientific Chemicals Supplier No. 4264103

Used in Ceramic tiles, white ware, glazes, glass, porcelain enamel frits, ceramics, filler, welding electrodes, and abrasives .

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula NaAlSi3O8
MDL Number MFCD00243144
Synonym Soda Spar

Specifications

Packaging Plastic bottle
Refractive Index 1.53
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 262.23
Chemical Name or Material Albite
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.