Learn More
Diagnostic Biosystems ALBUMIN Albumin
Supplier:  Diagnostic Biosystems RP04601
Encompass_Preferred
This antibody reacts with human albumin. This antibody is essentially free of other rabbit serum proteins. Specificity is determined by Ouchterlony double diffusion (ODD) and immunoelectrophoresis (IEP) versus human serum and human albumin.
Catalog No. 50-238-5429
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.