Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Albumin, Bovine, For IVD, Low Ash and Metals

Catalog No. ICN16006925 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN16006925 25 g
ICN16006910 10 g
ICN16006991 1 kg
ICN16006980 100 g
ICN16006990 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN16006925 Supplier MP Biomedicals™ Supplier No. 0216006925
Add to Cart
Add to Cart

Prepared by heat shock fractionation

  • For IVD applications
  • Low visible absorbance
  • Purity: >98%
  • Ash: <2%
  • Metals: <20ppm

For best cell culture performance, see alternate Mfr. No. 180577

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA, Microbial Culture
Molecular Weight (g/mol) 66.338
Name Bovine Albumin Fraction V
pH Range 6.5 to 7.5
Quantity 25 g
Source Bovine
Storage Requirements 2° to 8°C
Form Powder
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.