Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Albumin Bovine, Fraction V from Bovine

Catalog No. MP216006950 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP216006950 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP216006950 Supplier MP Biomedicals™ Supplier No. 0216006950
Add to Cart
Add to Cart

Albumin Bovine, Fraction V from Bovine≥98%, Off-white Powder

  • Albumin is general blocking reagent in ELISA, as a nutrient in cell & microbial culture, it also stabilize some enzymes during digestion of DNA.
  • It also used to determine the quantity of other proteins, by comparing an unknown quantity of protein to known amounts of BSA.
  • MP offers this grade of albumin as a more highly purified product than our traditional True Cohn Fraction V Powder.
  • Due to lower manufacturing costs, this grade is available at a substantial savings relative to our True Cohn Fraction V Powder.
TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) ∽66338
pH Range 6.5-7.5 (1% isotonic)
Quantity 50 g
Source Bovine
Storage Requirements +4°C
Product Type Proteins, Enzymes & Peptides
Form Powder
Purity or Quality Grade ≥98%
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.