Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Albumin, Bovine Fraction V, fatty acid free, low endotoxin, cell culture reagent

Catalog No. MP219477201 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219477201 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219477201 Supplier MP Biomedicals™ Supplier No. 0219477201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Albumin, Bovine Fraction V, fatty acid free, low endotoxin, cell culture reagent, Off White Shiny Flakes

  • Fraction V
  • Cell Culture Reagent
  • Fatty Acid Free
  • Low endotoxin
  • Albumins are a group of acidic proteins which occur in the body fluids and tissues of mammals and in some plant seeds.
  • Serum and plasma albumin is carbohydrate free and comprises 55-62% of the protein present. However, only about 40% of the total albumin in the body is in the circulating plasma at one time with the remainder being in extracellular spaces with which there is, in general, equilibration about every 24 hours.
TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) ∽66338
pH Range 6.8-7.2 (7% aq soln)
Quantity 1g
Storage Requirements +4°C.
Product Type Proteins, Enzymes and Peptides
Form Flakes
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.