Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Albumin, Chicken, MP Biomedicals™

Catalog No. MP219122425 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219122425 25 g
MP219122405 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP219122425 Supplier MP Biomedicals™ Supplier No. 0219122425
Add to Cart
Add to Cart

Useful as a marker for protein electrophoresis. Albumin, Chicken, MP Biomedicals, a phosphorylated glycoprotein, is the major protein constituent of egg whites. Ovalbumin can be used as a carrier protein to conjugate to synthetic peptides for use as an im

From the amino acid sequence, the peptide portion of the molecule consists of 385 residues and has a molecular weight of 42.7 kDa. Differential scanning calorimetry indicates that chicken egg albumin denatures at 84°C.

  • Can be used as a carrier protein to conjugate to synthetic peptides for use as an immunogen
  • Useful as a marker (∼45 kDa) for protein electrophoresis
  • Has been used to produce an animal model of asthma
  • Used as a medium for cell incubation in EliCell assay for the detection of released cytokines from eosinophil
  • Was used in a study testing intestinal macromolecule absorption in the fetal pig after infusion of colostrum in utero
TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 g
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.