Learn More
Supelco Inc ALBUMIN FROM BOVINE SERUM 25G
Supplier:  Supelco Inc A147025G
Encompass
Albumin, low endotoxin, lyophilized powder, Cohn AnAlogA„c, BioReAgent, suitAble for cell culture, A%Y98% (AgArose gel electrophoresis)
Catalog No. NC0354908
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.