Learn More
MP Biomedicals™ Albumin, Human, MP Biomedicals™
Catalog No. MP021913492 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021913492 250 mg
MP021913491 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP021913492 Supplier MP Biomedicals™ Supplier No. 02191349.2
Add to Cart
Add to Cart
Regulates colloidal osmotic blood pressure. Albumin, Human, MP Biomedicals functions as a carrier protein for steroids, fatty acids and thyroid hormones. Unlike globulins, albumins have low molecular weights, are soluble in water, are easily crystallized
Albumins are a group of simple proteins found in the body fluids and tissues of animals and in some plant seeds. Serum and plasma albumin is carbohydrate-free and comprises 55-62% of the protein present. Structurally it is a single polypeptide chain with one free sulfhydryl group on residue no. 34, plus 17 intrachain disulfide bonds. Due to its high charge to mass ratio, albumin binds water, Ca2+, Na+, K+, fatty acids, bilirubin, hormones and drugs. Albumin is also seen to bind to exogenous substances, particularly drugs (e.g., ibuprofen, warfarin), and strongly influence their pharmacokinetics. Oxidative stress leading to changes in the redox state of albumin has widely varied effects on its physiological function.
The main biological function of albumin is to regulate the colloidal osmotic pressure of blood. Human and bovine albumins contain 16% nitrogen and are often used as standards in protein calibration studies. Due to their free hydrophobic region, fatty acid free albumins are used to solubilize lipids in tissue culture. They are also used as blocking agents in Western blots or ELISA applications. Globulin-free albumins are suitable for use in applications where no other proteins should be present (e.g., electrophoresis).
TRUSTED_SUSTAINABILITY
Specifications
Quantity 250 mg
WARNING: Cancer - www.P65Warnings.ca.gov