Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Abnova™ Albumin (Mouse) ELISA Kit

Catalog No. 89100235 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-235 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-235 Supplier Abnova™ Supplier No. KA0488
Add to Cart
Add to Cart

Albumin (Mouse) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of mouse albumin in plasma and serum.

  • Designed for detection of albumin in mouse plasma and serum samples
  • Employs a quantitative competitive enzyme immunoassay technique that measures mouse albumin in approximately 2 hours
  • A polyclonal antibody specific for mouse albumin has been pre-coated onto a 96-well microplate with removable strips
  • Upon arrival, immediately store components of the kit at recommended temperatures up to the expiration date

Specifications

Calibration Range 0.098 to 25μg/mL
Description Albumin (Mouse) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of mouse albumin in plasma and serum.
Detection Method Colorimetric
Product Type Albumin (Mouse) ELISA Kit
Sample Type Plasma, Serum
Target Species Mouse
For Use With (Application) Quantification
Gene ID (Entrez) 11657
Gene Symbol Gene Alias: Alb-1, Alb1
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 25 μL
Storage Requirements Store components of the kit at 4°C or -20°C as described in the protocol.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.