Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Abnova™ Albumin (Mouse) ELISA Kit

Catalog No. 89100234 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-234 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-234 Supplier Abnova™ Supplier No. KA0489
Add to Cart
Add to Cart

Albumin (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse albumin in cell culture supernatant and urine.

  • Designed for detection of mouse albumin in urine and cell culture samples
  • Employs a quantitative sandwich enzyme immunoassay technique that measures mouse albumin in less than 4 hours
  • A polyclonal antibody specific for mouse albumin has been pre-coated onto a 96-well microplate with removable strips
  • Upon arrival, immediately store components of the kit at recommended temperatures up to the expiration date

Specifications

Assay Range Intra-Assay: 4.50%, Inter-Assay: 7.40%
Calibration Range 1.56 to 400ng/mL
Description Albumin (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse albumin in cell culture supernatant and urine.
Detection Method Colorimetric
Product Type Albumin (Mouse) ELISA Kit
Sample Type Cell Culture Supernatant, Urine
Target Species Mouse
For Use With (Application) Quantification
Gene ID (Entrez) 11657
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 50 μL
Storage Requirements Store components of the kit at 4°C or -20°C as described in the protocol.
Synonym Gene Alias: Alb-1, Alb1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.