Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Albumin Native Protein
SDP

Catalog No. NB816197 Shop All Novus Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB816197 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB816197 Supplier R&D Systems™ Supplier No. NBP1970495MG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Native Protein

Specifications

Accession Number P02768
For Use With (Application) Dot Blot, PAGE
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Gene ID (Entrez) 213
Quantity 5 mg
Storage Requirements Store at 4°C.
Gene Alias albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin
Gene Symbol ALB
Reconstitution Reconstitute with 500 μL of deionzied water.
Inhibitor Properties This product has been tested in SDS-Page and can be used in SDS, Western Blotting, ELISA experiments and other immunological assays.
Research Category Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Stem Cell Markers
Product Line Novus
Purity or Quality Grade Multi-step
Protein Albumin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.