Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ AlbumiNZ™ Ultra-Low IgG BSA

Catalog No. 02FC007650 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02FC007650 50 g
02FC007610 10 g
02FC007680 100 g
02FC007690 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02FC007650 Supplier MP Biomedicals™ Supplier No. 02FC007650

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Manufactured in a state-of-the-art facility in New Zealand using a chromatographic extraction process under Quality Systems that are ISO Certified and audited to cGMP Principles.

  • While the low IgG BSA (Cat.#02199897) has an IgG limit of up to 50 ppm, in certain applications this level becomes an antigen for cross-reacting secondary antibodies
  • Its presence can alter precious experimental data, resulting in reduced purification yields
  • These levels may also be problematic for certain recombinant protein or monoclonal antibody purification processes
  • For such applications, MP Bio NZ offers an ultra-low IgG grade of BSA where the IgG content is brought down to as low as 4 ppm
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Refrigerate
Type Ultra-Low IgG BSA
Product Line AlbumiNZ
Quantity 50 g
Species Bovine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.