Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcian Blue 8GX, Sigma-Aldrich

Catalog No. 01259062 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-062 25 g
01-259-089 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-259-062 Supplier MilliporeSigma Supelco Supplier No. A526825G
Add to Cart
Add to Cart

Alcian Blue 8GX, also known as Ingrain Blue 1, is a basic copper phthalocyanine dye whose copper content imparts a blue color. It is tetravalent, cationic, and water-soluble in nature. It forms salt linkages with the acid groups of acid mucopolysaccharides.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Alcian Blue
CAS 33864-99-2
Molecular Weight (g/mol) 1298.86 g/mol
Color Blue
Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.