Learn More
StatLab ALCIAN BLUE PH 2.5 100ML
SDP
Supplier:  StatLab STABL2.5100
Encompass_Preferred
Stain; Alcian Blue; 2.5 pH; 100mL; Stains for sulfated mucosubstances
Catalog No. NC9070091
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.