Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Alcian Blue (pH 2.5) Stain Kit
SDP

Catalog No. NB398957 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398957 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398957 Supplier R&D Systems™ Supplier No. H3501NB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Intended for use in the histological visualization of sulfated and carboxylated acid mucopolysaccharides and sulfated and carboxylated sialomucins (glycoproteins).intended for use in the histological visualization of sulfated and carboxylated acid mucopolysaccharides and sulfated and carboxylated sialomucins (glycoproteins).

  • Staining Results-
    • Acidic Sulfated Mucosubstances-Blue
    • Hyaluronic Acid-Blue
    • Sialomucins-Blue
    • Nuclei-Red
  • Control Tissue- Small Intestine, Appendix, Colon
  • Components-
    • Acetic Acid Solution (3%)- 2 x 30 mL
    • Alcian Blue Solution (pH 2.5)- 30 mL
    • Nuclear Fast Red (Enhanced)- 30 mL
  • Storage-Room temperature
  • Guarantee-6 months from date of receipt

Specifications

Product Line Novus
Quantity 1 Kit
Storage Requirements Room temperature
Product Type Stain Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.