Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol, 190 Proof, USP, 94.9-96%, Spectrum™ Chemical
SDP

Catalog No. 18600371 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
4 x 4 L
55 gal.
Packaging:
Amber Glass Bottle
Drum
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-371 4 L Amber Glass Bottle
18-600-373 4 x 4 L Amber Glass Bottle
18-600-374 55 gal. Drum
18-600-367 100 mL Amber Glass Bottle
18-600-368 500 mL Amber Glass Bottle
18-600-370 1 L Amber Glass Bottle
18-600-369 6 x 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-600-371 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ET1084LTGL
Add to Cart
Add to Cart

ET108, 64-17-5, C2H5OH

Spectrum™ Chemical Alcohol, 190 Proof, USP is approximately 95% Ethanol and 5% water. This product, 190 proof USP grade alcohol, is used as an antiseptic to disinfect skin prior to injections and as a solvent. All Spectrum Chemical USP products, including all USP alcohol products, are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

CAS Max % 100%
Quantity 4 L
Packaging Amber Glass Bottle
Specific Gravity 0.812-0.816
UN Number UN1170
Concentration 95%
Boiling Point 79°C
Percent Purity 94.9 to 96%
Grade USP
Chemical Name or Material Alcohol
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.