Learn More
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 66.39, 30.25, 1.68, 1.68 |
| Decomposition Information | Carbon monoxide Carbon dioxide May release flammable gases |
| Quantity | 125 mL |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Physical Form | Liquid |
| Linear Formula | HOCH2CH(OH)CH2OH |
| UN Number | UN1987 |
| Solubility Information | Soluble in water |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Flammable liquid and vapor. Causes skin and serious eye irritation. May cause drowsiness or dizziness. Suspected of damaging fertility or the unborn child. Causes damage to organs (central nervous system, optic nerve). |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Keep away from heat, hot surfaces, open flames, sparks. Keep container tightly closed. Ground/bond container and receiving equipment. Use explosion-proof electrical, lighting, ventilating equipment. Use only non-sparking tools. Take precautionary measures against static discharge. Do not breathe mist, spray, vapors. Use only outdoors or in a well-ventilated area. Do not eat, drink or smoke when using this product. Wear eye protection, face protection, protective clothing, protective gloves. Wash exposed skin thoroughly after handling. If exposed or concerned: Get medical advice/attention. If on skin (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. If skin irritation occurs: Get medical advice/attention. Wash contaminated clothing before reuse. If inhaled: Remove person to fresh air and keep comfortable for breathing. Call a poison center/doctor if you feel unwell. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical attention. In case of fire: Use dry chemical powder, alcohol-resistant foam, carbon dioxide to extinguish. Store locked up in a cool, well-ventilated place. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.