Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Alcohol Prep Pads

Catalog No. 19090679 Shop All Medique Products
Change view
Click to view available options
Quantity:
10 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-679 10 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-090-679 Supplier Medique Supplier No. 22112
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Prevents infection in minor cuts, scrapes and abrasions.

Contains 70% isopropyl

Specifications

Active Ingredients Isopropyl alcohol 70%
Product Type Alcohol Prep Pads
For Use With (Application) Prevents infection in minor cuts, scrapes and abrasions
Quantity 10 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.