Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol, Reagent, 20% (v/v), Ricca Chemical™
SDP

Catalog No. SA93201 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SA9320-1 1 L Natural Poly Bottle
SA9320-4 4 L Natural Poly Bottle
SA932010 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SA9320-1

Inactive Item. Please call Customer Service Representative.

Chemical Identifiers

CAS 7732-18-5 , 64-17-5 , 67-63-0 , 67-56-1
Molecular Formula C2H6O
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

CAS Min % 79.0, 18.0, 0.8, 0.9
CAS Max % 81.0, 20.0, 0.9, 1.0
Quantity 1 L
Packaging Natural Poly Bottle
Boiling Point <100°C
Molecular Weight (g/mol) Mixture
Percent Purity 20% v/v
Grade Laboratory
Chemical Name or Material Reagent Alcohol, 20% (v/v)
Safety handling Check Alert storage code Red
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.