Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcohol SDA-3A 200 Proof, Solstice

Catalog No. 60090196 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1250 L
Packaging:
IBC
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-090-196 1250 L IBC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-196 Supplier Solstice Supplier No. CS094ZA1250

missing translation for 'validMainframeMsg'

Specially denatured alcohol, contains 5% methanol, 200 proof

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5 , 67-56-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

CAS Max % 5.00%, 95.00%
Quantity 1250 L
Flash Point 15°C.The physical data is that of the main component.
Packaging IBC
Physical Form Liquid
Boiling Point 78.32°C.The physical data is that of the main component.
Grade Reagent grade solvents
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.