Learn More
TARGETMOL CHEMICALS INC ALDA-1 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2662
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Alda-1 is an ALDH2 agonist a cell-permeable activator of both the wild-type ALDH2*1 and the Asian E487K mutant ALDH2*2 forms of mitochondrial aldehyde dehydrogenase 2 (mtALDH2). purity: 99%
Catalog No. 50-258-0685
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.