Learn More
WCM INC Aldex GL1 8oz 48/CS, 8 oz treats 1G, total 48 Gallons
Supplier:  WCM INC AMSGL1
Encompass_Preferred
Aldex GL1, 8oz bottles, 48 bottles total. 8 oz liquid treats 1 Gallon. Neutralizer converts liquid aldehyde waste (10% Formalin, 4% Glutaraldehydes or 3.75% Formaldehydes) into a non hazardous liquid safe for drain disposal. Treats a total of 48 Gallons
Catalog No. NC9127919
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.