Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Aldh Activity Assay Kit, Fluorometric, Quantitative, 100 Tests

Catalog No. 502582881
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-2881 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-2881 Supplier Abcam Supplier No. AB155894100TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorometric assay kit for sensitive and quantitative measurement of aldehyde dehydrogenase activity in various biological samples.

  • Assay type: Enzyme activity
  • Detection method: Fluorescent
  • Sensitivity: < 0.05 mU/well
  • Results type: Quantitative
  • Sample types: Plasma, Cell culture supernatant, Serum, Cell Lysate, Tissue Culture Media
  • Applications: Enzyme activity assay
  • Cited in: 3 publications
  • Compatible hardware: Microplate reader

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Aldehyde Dehydrogenase
Format Kit
For Use With (Equipment) Microplate Reader
No. of Reactions 100 Tests
Detection Method Fluorescence
Sample Type Plasma, Cell culture supernatant, Serum, Cell lysate, Tissue culture media
Quantity 100 Tests
For Use With (Application) Enzyme activity assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.