Learn More
Cell Signaling Technology ALDH1A1 D4R9V Rabbit Mab, Monoclonal Antibody, Recombinant Production, 100 µl
SDP
Supplier:  Cell Signaling Technology 12035S
Encompass
ALDH1A1 (D4R9V) Rabbit Monoclonal Antibody is a highly specific antibody for detecting ALDH1A1, crucial in alcohol and retinol metabolism, and a potential cancer stem cell biomarker. Its recombinant production ensures superior lot-to-lot consistency, offering an animal-free supply. Validated for use in Western Blot and Immunoprecipitation, it is effective with human and mouse samples, making it a reliable choice for researchers in various malignancies.
Catalog No. NC2259523
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.