Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH1A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595937
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595937 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595937 Supplier Invitrogen™ Supplier No. PA595937
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: A-549, HT-29, K-562, Mouse liver, Mouse lung, Rat lung; Cellular Location: Cytoplasm Immunogen sequence: KYILGNPLTP GVTQGPQ.

ALDH1A1 belongs to the aldehyde dehydrogenases family of proteins. Aldehyde dehydrogenase is the second enzyme of the major oxidative pathway of alcohol metabolism. Two major liver isoforms of this enzyme, cytosolic and mitochondrial, can be distinguished by their electrophoretic mobilities, kinetic properties, and subcellular localizations. Most Caucasians have two major isozymes, while approximately 50% of Orientals have only the cytosolic isozyme, missing the mitochondrial isozyme. A remarkably higher frequency of acute alcohol intoxication among Orientals than among Caucasians could be related to the absence of the mitochondrial isozyme.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH1A1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ALDH1A1
Gene Accession No. P00352, P24549, P51647
Gene Alias acetaldehyde dehydrogenase 1; Ahd2; Ahd-2; alcohol dehydrogenase family 1, subfamily A1; alcohol dehydrogenase family 1, subfamily A2; ALDC; aldehyde dehydrogenase 1 family member A1; aldehyde dehydrogenase 1 family, member A1; Aldehyde dehydrogenase 1 soluble; aldehyde dehydrogenase 1, liver cytosolic (class 1); aldehyde dehydrogenase 1, soluble; aldehyde dehydrogenase 1, subfamily A1; aldehyde dehydrogenase 1A1; aldehyde dehydrogenase 2; aldehyde dehydrogenase family 1 member A1; aldehyde dehydrogenase family 1, member A1; aldehyde dehydrogenase family 1, subfamily A1; aldehyde dehydrogenase, cytosolic; aldehyde dehydrogenase, liver cytosolic; Aldh; ALDH class 1; Aldh1; ALDH11; ALDH1A1; Aldh1a2; Aldh2; ALDH-E1; ALHDII; E1; epididymis luminal protein 12; epididymis luminal protein 9; epididymis secretory sperm binding protein Li 53e; epididymis secretory sperm binding protein Li53e; HEL12; HEL-9; HEL-S-53e; MGC2318; PUMB1; RALDH 1; ralDH1; Retinal dehydrogenase 1; retinaldehyde dehydrogenase 1
Gene Symbols ALDH1A1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 301-400 of human ALDH1A1 (NP_0006802).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11668, 216, 24188
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.