Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALDH1A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522377
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522377 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522377 Supplier Invitrogen™ Supplier No. PA522377
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: IMR32, mouse liver, mouse testis, rat testis. Predicted reactivity: Mouse (97%), Rat (98%), Xenopus laevis (91%), Dog (99%), Chicken (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This protein belongs to the aldehyde dehydrogenase family of proteins. The product of this gene is an enzyme that catalyzes the synthesis of retinoic acid (RA) from retinaldehyde. Retinoic acid, the active derivative of vitamin A (retinol), is a hormonal signaling molecule that functions in developing and adult tissues. The studies of a similar mouse gene suggest that this enzyme and the cytochrome CYP26A1, concurrently establish local embryonic retinoic acid levels which facilitate posterior organ development and prevent spina bifida. Three transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALDH1A2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene ALDH1A2
Gene Accession No. O94788, Q62148, Q63639
Gene Alias AL1A2; alcohol dehydrogenase family 1, subfamily A2; alcohol dehydrogenase family 1, subfamily A7; aldehyde dehydrogenase 1 family member A2; aldehyde dehydrogenase 1 family, member A2; aldehyde dehydrogenase 1A2; aldehyde dehydrogenase family 1 member A2; aldehyde dehydrogenase family 1, subfamily A2; Aldh1a2; Aldh1a7; AV116159; fb50h01; neckless; nls; nof; no-fin; RALDH 2; RALDH(II); Raldh1; RALDH2; Raldh-2; RALDH2-T; retinal dehydrogenase 2; retinal dehydrogenase, type II; retinaldehyde dehydrogenase 2; retinaldehyde dehydrogenase type 2; retinaldehyde-specific dehydrogenase type 2
Gene Symbols ALDH1A2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ALDH1A2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116676, 19378, 8854
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.