Learn More
Description
Positive Control: A431, Huh-7, U87-MG, SK-N-AS Predicted Reactivity: Mouse (95%), Rat (95%), Xenopus laevis (83%), Chicken (88%), Rhesus Monkey (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | ALDH1A3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | GT926 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | ALDH1A3 |
| Gene Accession No. | P47895, Q8K4D8, Q9JHW9 |
| Gene Alias | acetaldehyde dehydrogenase 6; aldehyde dehydrogenase 1 family member A3; aldehyde dehydrogenase 1 family, member A3; aldehyde dehydrogenase 6; Aldehyde dehydrogenase family 1 member A3; aldehyde dehydrogenase family 1, subfamily A3; aldehyde dehydrogenase-6; ALDH1A3; ALDH1A6; ALDH6; ALDHA1A3; MCOP8; ralDH3; RALDH-3; Retinaldehyde dehydrogenase 3; V1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
