Learn More
Invitrogen™ ALDH1L1 Monoclonal Antibody (2E7)

Description
ALDH1L1 Monoclonal Antibody for Western Blot, ICC/IF
Specifications
Specifications
| Antigen | ALDH1L1 |
| Applications | Western Blot, Immunocytochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 2E7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 5mM sodium azide |
| Gene | ALDH1L1 |
| Gene Accession No. | O75891, P28037, Q8R0Y6 |
| Gene Alias | 10-formyltetrahydrofolate dehydrogenase; 10fTHF; 10-fTHF; 10FTHFDH; 10-FTHFDH; 1810048F20Rik; aldehyde dehydrogenase 1 family member L1; aldehyde dehydrogenase 1 family, member L1; aldehyde dehydrogenase family 1 member L1; Aldh1l1; Cytosolic 10-formyltetrahydrofolate dehydrogenase; DKFZp781N0997; FBP-CI; FDH; formyltetrahydrofolate dehydrogenase; Fthfd; LOC100855730; MER57B-ALDH1L1; Neut2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.