Learn More
WCM INC Aldex AMS3010, Crystals. Each container treats 7.5 Gallons of Aldehyde waste.
Supplier:  WCM INC AMS3010
Encompass_Preferred
Each bottle treats 7.5 gallons of aldehyde.Aldex AMS3010 crystals is perfect for your formalin, aldehyde disposal. Add crystals to containers holding solution and dispose in the trash. Aldex neutralizes and crosslink¿s 10% formalin or up to 4% glutaraldehyde or 3.75% Glutaraldehye and converts it into a non-toxic, non-hazardous end-product that can be safely disposed in your regular trash. Aldex also reduces exposure to potentially hazardous vapor emanating from most aldehydes.
Catalog No. NC9126907
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.