Learn More
Cayman Chemical GonadorelInacetate 25mg
SDP
Supplier:  Cayman Chemical 2424025 MG
Encompass_Preferred
A PAD4 inhibitor (IC50 2.3 UM) selective for PAD4 over PAD1 PAD2 and PAD3 (IC50s 8.5 71 and 26 UM respectively) reduces global citrullination in HL-60 granulocytes at 1 10 and 100 nM reduces citrullination of H3R2Cit/R8Cit/R17Cit in MCF-7 cells at 1 and 10 UM
Catalog No. 50-293-5644
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.