Learn More
MilliporeSigma™ Supelco™ Aldrich™ TLC Developing Tank - Cylindrical Tank Lid
Catalog No. 111011786 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
Glass
Catalog No. Material
11-101-1786 Glass
1 options
Catalog No. 11-101-1786 Supplier MilliporeSigma™ Supelco™ Supplier No. Z4072591EA
Add to Cart
Add to Cart
Aldrich™ TLC developing tanks are used to develop TLC plates.
Can be used as a staining chamber. Made of glass with lid. Compact size offers good plate visibility and minimal solvent use.
TRUSTED_SUSTAINABILITY
Specifications
Material Glass
Quantity 1/Pk.
For Use With (Application) TLC
For Use With (Equipment) Aldrich™ TLC Developing Tank - Cylindrical Glass Tank
Product Line Aldrich
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC