Learn More
Sigma Aldrich Fine Chemicals Biosciences DuranR graduated laborator
Supplier: Sigma Aldrich Fine Chemicals Biosciences Z23220310EA
Reusable bottles without caps or pouring rings for the storage of reagents chemicals SOLUTIONons tissue culture media etc. Made of Duran borosilicate glass Type 3.3 for maximum chemical and thermal shock resistance.Optional pouring rings prevent drips when pouring from these bottles.