Learn More
Abbott™ Cholestech LDX™ Calibration Verification Materials, Levels 1 through 4
Catalog No. 23763106 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
4 x 2 mL
Catalog No. Quantity
23-763-106 4 x 2 mL
1 options
This item is not returnable. View return policy
Use universal precautions. All human source material used to manufacture this product was nonreactive for HBsAg, HIV-1/HIV-2, HCV, HIV-1 RNA, HCV RNA and nonreactive to STS using U.S. FDA-specified techniques, but complete absence of human pathogens is not guaranteed.
Catalog No. 23-763-106 Supplier Abbott™ Supplier No. 88770
Add to Cart
Add to Cart
This item is not returnable. View return policy
Use universal precautions. All human source material used to manufacture this product was nonreactive for HBsAg, HIV-1/HIV-2, HCV, HIV-1 RNA, HCV RNA and nonreactive to STS using U.S. FDA-specified techniques, but complete absence of human pathogens is not guaranteed.
Designed for use in verifying the reportable range of tests on the Cholestech™ LDX system
  • Analytes in the calibration materials include total cholesterol, HDL cholesterol, triglycerides and glucose
  • Compare results obtained with assigned ranges given in the package insert
Specifications
Type Cholestech Calibration Verification Material
Intended Use Verifying reportable ranges of total Cholesterol, HDL, Triglycerides, Glucose
Level 1 to 4
Quantity 4 x 2 mL