Learn More
Abbott™ Cholestech LDX™ Multianalyte Controls
Catalog No. 23763176 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
1 x 0.25 mL
2 x 2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-763-176 1 x 0.25 mL
23-763-107 2 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Human and animal source material: treat as potentially infectious. Do not use reagents after expiration date printed on vial labels.
Catalog No. 23-763-176 Supplier Abbott™ Supplier No. 88773
Add to Cart
Add to Cart
This item is not returnable. View return policy
Human and animal source material: treat as potentially infectious. Do not use reagents after expiration date printed on vial labels.
Enables users to monitor the performance of the Cholestech LDX system for commonly tested analytes.
  • Analytes in these controls include total cholesterol (TC), high density lipoprotein (HDL), triglycerides (TRG), glucose (GLU), alanine aminotransferase (ALT), and aspartate aminotransferase (AST)
  • Reagents are prepared from human and animal constituents in an aqueous preservative medium containing antimicrobial agents
Specifications
Type Cholestech LDX Multianalyte Control
Level 1, 2
Quantity 1 x 0.25 mL