Learn More
C. DIFF QUIK CHEK COMPLETE™
GSA_VA
Catalog No. 23900551
Change view
Click to view available options
Quantity:
25 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-900-551 50 Tests
23-900-552 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-900-551 Supplier Techlab Inc Supplier No. 30550C
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
The C. DIFF QUIK CHEK COMPLETE™ test is a rapid membrane enzyme immunoassay for the simultaneous detection of Clostridioides difficile glutamate dehydrogenase antigen and toxins A and B in a single reaction well. Developed and Manufactured by TECHLAB, Inc.
  • Results in approximately 30 minutes
  • Easy to intrepret
  • Highly sensitive and specific
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Detectable Analytes C. difficile GDH Antigen, Toxins A and B
Type Clostridium dificile QUIK CHEK COMPLETE™
Includes 25 Membrane device pouches, diluent, wash buffer, conjugate, substrate, positive control, disposable plastic transfer pipettes
Quantity 50 Tests