Learn More
Description
Transforming near-patient testing
- Unique isothermal amplification technology giving you molecular results, faster than ever before
- Giving you the confidence to make effective clinical decisions sooner
- Enables prompt initiation of infection control measures
- Connected results direct to your network
- Facilitating effective patient management
Enabling effective Antimicrobial Stewardship
- Molecular sensitivity in an actionable timeframe allows the prudent use of antibiotics and antivirals
- Reliable near-patient testing reduces overall healthcare costs
- Intuitive and reliable, allowing you to respond quickly and accurately in any setting
- Simple and easy to use molecular diagnostic test
- Sample type nasal swab
- Requires only minimal training
- Small footprint can be used in any healthcare setting
- Streamlined workflow connectivity can be provided to your network
- Demonstrates impressive sensitivity for the detection of both influenza A and B in clinical respiratory specimens
Order Info
See ID NOW Influenza A and B Control Swab Kit (23-046-619)
Specifications
Specifications
| Product Type | Abbott ID NOW Instrument |
| For Use With (Application) | Infectious disease diagnosis |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.