Learn More
Abbott™ iCup™ A.D. Zero Exposure Urine Drug Screens
Description
Detect the presence of drugs and adulterants in just minutes
- The iCup AD Drug Screen offers versatility with up to 12 drug assays in one cup
- Multiple configurations available
- Built-in validity screening to alleviate tampering concerns
- Room temperature storage
- Comes with temperature strip to verify range of collected sample
- Results can be photocopied for record keeping purposes
- 24-month shelf life
Specifications
Specifications
| Detectable Analytes | Cocaine (COC), Cannibinoids (THC), Opiates (OPI), Amphetamine (AMP), Methamphetamine (mAMP), Benzodiazepines (BZO), Barbiturates (BAR), Oxycodone (OXY), Methodone (MTD), Propoxyphene (PPX) |
| Sample Type | Urine |
| Type | iCup™ A.D. Urine Drug Screen |
| Quantity | 25 Pack |