Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Triage™ BNP Test for Beckman Coulter Immunoassay Systems
SDP

Catalog No. 1398201
Change view
Click to view available options
Quantity:
1 Pack
100 Pack
Type:
Triage™ BNP Calibration/Verification Test
Triage™ BNP Control Kit
Triage™ BNP Test Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
13-982-01 1 Pack Triage™ BNP Control Kit
13-982-00 100 Pack Triage™ BNP Test Kit
13-982-02 1 Pack Triage™ BNP Calibration/Verification Test
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 13-982-01 Supplier Quidel Supplier No. 98201
This item is not returnable. View return policy

Empowers higher throughput BNP with rapid testing and automation

Provides clinically interchangable BNP results between Alere Triage™ MeterPro and Beckman Coulter UniCel DxI 800 or Beckman Coulter Access 2 Immunoassay System

  • Yields results in 15 minutes
  • Uses plasma sample
  • Aids in the diagnosis and severity of CHF
  • Risk stratifies patients with HF and ACS
  • Consistent BNP results in emergency department, point of care or labs

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 10°C
Type Triage™ BNP Control Kit
For Use With (Application) Diagnosis of congestive heart failure
Quantity 1 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.