Learn More
Abbott™ Impact RPR™ Test Kits
Catalog No. WA60C2 Shop All Abbott Rapid Diagnostics Products
Change view
Click to view available options
Quantity:
150 Tests
50 Tests
500 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
WA-60C2 150 Tests
WA-60C1 50 Tests
WA-60C3 500 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
RPR Card Antigen Suspension contains sodium azide, which is harmful if swallowed. Contact with acids liberates very toxic gas. After contact with skin, wash immediately with plenty of water. Sodium azide may react with lead and copper plumbing to form highly explosive metal azides. On disposal, flush with a large volume of water to prevent azide buildup.
Catalog No. WA-60C2 Supplier Abbott™ Supplier No. 60C2
Add to Cart
Add to Cart
This item is not returnable. View return policy
RPR Card Antigen Suspension contains sodium azide, which is harmful if swallowed. Contact with acids liberates very toxic gas. After contact with skin, wash immediately with plenty of water. Sodium azide may react with lead and copper plumbing to form highly explosive metal azides. On disposal, flush with a large volume of water to prevent azide buildup.
Includes
RPR Test Kits: Antigen suspension, dispensing bottle with needle and sheath, pipet/stirrers, and plastic test cards with ten or thirty 18mm diameter wells that will not warp or bow
IMPACT RPR™ Card Test is a non-treponemal test for the rapid detection and quantitative determination of syphilis in serum or plasma.
  • Classified as a standard test for syphilis by the Centers for Disease Control and Prevention (CDC)
  • Non-treponemal test for the serological detection of syphilis
  • Assay utilizes carbon particle technology
Order Info
Controls available separately in both dry and liquid form.
Specifications
Detectable Analytes Reagin (Syphilis)
Type RPR Test Kit
Includes Antigen suspension, dispensing bottle with needle and sheath, pipet/stirrers, Plastic test cards with ten 18mm diameter wells
Quantity 150 Tests